Learn More
Electron Microscopy SciencesSupplier Diversity Partner Light Box, Model A  DFS Item
Manufacturer:  Electron Microscopy SciencesSupplier Diversity Partner 71650
For viewing and editing slides, transparencies, negatives, micrographs, etc. They also serve as a base for orientation of embedding specimens when observing tissues using a magnifier. Scientifically designed to provide even distribution and uniformity of light. All metal welded construction with a white baked-enamel finish. The viewing surface is a translucent acrylic that is break resistant. It can be used flat on a table, inclined, standing, or hung on the wall. It comes with a ANSI standard 5000K fluorescent lamp, 10 feet cord, on-off switch. It is compact and can be used on a desk. Small enough to carry in a briefcase or store in a drawer. Viewing surface 8 3/4"x11.5" (22x29.2cm). Overall: 9.5x12.5x2"H.
Catalog No. 50-949-588