Learn More
Kemtec™ Light & Color Kit
Kemtec™ Light & Color Kit includes fourteen activities and four demonstrations to introduce students to the properties of light
Manufacturer: Kemtec™ 1122
Description
- Kit includes fourteen activities and four demonstrations
- Introduce properties of waves, properties of light, and the electromagnetic spectrum
- Includes reflection of light, pool reflections, reflection from plane mirrors, reflection from concave and convex mirrors, refraction through concave and convex lenses
- Features using a prism to separate white light into the colors of the visible spectrum
- Teaches how reflection determines color, mixing primary colors of light, and mixing the primary colors of pigments
- Complete with all materials for station based investigation, reproducible student data sheets, assessment questions, and answer keys
- Addresses national standards for Grades 5-8
Specifications
5 to 8 | |
Grades 5-8 | |
Light & Color | |
12 Lab Periods | |
19 x 12 x 5 in. |
10 to 13 | |
Instructor's Manual, Colored Filters, Convex/Concave Mirrors and Lenses, Prism, Flashlights (6), Paint, Colored Pencils, Toothpicks, Grating Cards, Index Cards, Cardboard, Light Box Template, Duct Tape, Straws, Binder Clips | |
1 Kit | |
5 lb. | |
24 Students |