Learn More
EVOS™ Light Cubes Available on GSA/VA Contract for Federal Government customers only.
Image fluorescent samples for specific wavelength
Manufacturer:  EVOS™ AMEP4668
 View more versions of this product
Catalog No. 12-563-571
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Light cube
TagBFP
Excitation/Emission: 390/18nm, 447/60nm
CascadeBlue?, Pacific Blue?, TagBFP and other fluorophores
EVOS™ FL Color Imaging System, FL Auto Imaging System, FL Imaging System, Countess™ II FL Automated Cell Counter