Learn More
EVOS™ Light Cubes
Image fluorescent samples for specific wavelength
Manufacturer:  EVOS™ AMEP4669
 View more versions of this product
Catalog No. 12-563-572
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Light cube
CFP-YFP
Excitation/Emission: 445/45nm, 542/27nm
CFP-YFP FRET emission and other fluorophores
EVOS™ FL Color Imaging System, FL Auto Imaging System, FL Imaging System, Countess™ II FL Automated Cell Counter