Learn More
EVOS™ Light Cubes Available on GSA/VA Contract for Federal Government customers only.
Image fluorescent samples for specific wavelength
Manufacturer:  EVOS™ AMEP4671
 View more versions of this product
Catalog No. 12-563-573
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Light cube
AO Red
Excitation/Emission: 442/46nm, 628/32nm
Acridine orange, CTC formazan, FuraRed? and other fluorophores
EVOS™ FL Color Imaging System, FL Auto Imaging System, FL Imaging System