Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Light green SF yellowish (C.I. 42095), For Microscopy, Certistain™, MilliporeSigma™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
5141-20-8
C37H34N2Na2O9S3
792.844
DGOBMKYRQHEFGQ-UHFFFAOYSA-L
C.I. 42095
21223
CHEBI:87065
disodium;3-[[N-ethyl-4-[[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]-(4-sulfonatophenyl)methyl]anilino]methyl]benzenesulfonate
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=C(C=C5)S(=O)(=O)[O-].[Na+].[Na+]
5141-20-8
C37H34N2Na2O9S3
792.844
DGOBMKYRQHEFGQ-UHFFFAOYSA-L
C.I. 42095
21223
CHEBI:87065
disodium;3-[[N-ethyl-4-[[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]-(4-sulfonatophenyl)methyl]anilino]methyl]benzenesulfonate
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=C(C=C5)S(=O)(=O)[O-].[Na+].[Na+]
For microscopy
Appearance Dark violet solid
Formula Weight 792.86
Physical Form Solid
Odor Weak odor
Packaging Glass bottle
Quantity 25g
Chemical Name or Material Light green SF yellowish