Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Light green SF yellowish (C.I. 42095), For Microscopy, Certistain™, MilliporeSigma™
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Description
For microscopy
Specifications
Formula Weight | 792.86 |
Odor | Weak odor |
Packaging | Glass bottle |
Color | Dark Violet |
Quantity | 25 g |
Physical Form | Solid |
Chemical Name or Material | Light green SF yellowish |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction