Novus Biologicals$trade; Lightning-Link Rapid Type A Biotin Antibody Labeling Kit
Label antibodies, proteins, peptides and other biomolecules with biotin, using simple procedure. Set up conjugations in seconds, and use them within minutes.
Manufacturer: Novus Biologicals$trade; 3700015
 View more versions of this product
Catalog No. 37-000-15
  N/A
Add to Cart
Alert
For Research Use Only
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Quantity 1 x 1-2mg Reaction
    Storage Requirements Store at −20°C. Avoid freeze-thaw cycles.
    Product Type Antibody Labeling Kit
    • No desalting or dialysis steps
    • Lightning-link Rapid technology can be used to label small quantities of antibody with 100% recovery
    • Type A biotin kit is intended for assays in which a streptavidin-labeled detection reagent will be used.
  • Description & Specifications
Related Products
Product Content Feedback