Novus Biologicals™ Lightning-Link Rapid Type A Biotin Antibody Labeling Kit
Label antibodies, proteins, peptides and other biomolecules with biotin, using simple procedure. Set up conjugations in seconds, and use them within minutes.
Manufacturer: Novus Biologicals™ 3700030
 View more versions of this product
Catalog No. 37-000-30
  Check Availability
Add to Cart
Alert
For Research Use Only
    Specifications
    Quantity 3 x 10-20μg Reaction
    Storage Requirements Store at −20°C. Avoid freeze-thaw cycles.
    Product Type Antibody Labeling Kit
    • No desalting or dialysis steps
    • Lightning-link Rapid technology can be used to label small quantities of antibody with 100% recovery
    • Type A biotin kit is intended for assays in which a streptavidin-labeled detection reagent will be used.
  • Description & Specifications
Related Products
Product Content Feedback