Novus Biologicals™ Lightning-Link Rapid Type B Biotin Antibody Labeling Kit
Label antibodies, proteins, peptides and other biomolecules with biotin, using simple procedure. Set up conjugations in seconds, and use them within minutes.
Manufacturer: Novus Biologicals™ 3710015
 View more versions of this product
Catalog No. 37-100-15
  Check Availability
Add to Cart
Alert
For Research Use Only
    Specifications
    Product Type Antibody Labeling Kit
    Quantity 1 x 1-2mg Reaction
    Storage Requirements Store at −20°C. Avoid freeze-thaw cycles.
    • No desalting or dialysis steps
    • Lightning-link Rapid technology can be used to label small quantities of antibody with 100% recovery
    • Type B biotin kit is intended for assays in which the biotinylated protein is captured by streptavidin immobilized on a surface (plates, nitrocellulose, magnetic beads, etc.)
  • Description & Specifications
Related Products
Product Content Feedback