Learn More
Novus Biologicals™ Lightning-Link Rapid Type B Biotin Antibody Labeling Kit
Label antibodies, proteins, peptides and other biomolecules with biotin, using simple procedure. Set up conjugations in seconds, and use them within minutes.
Manufacturer:  Novus Biologicals™ 3710030
 View more versions of this product
Catalog No. 37-100-30
  Check Availability
Add to cart
Alert:
For Research Use Only
Specifications
Description
  • No desalting or dialysis steps
  • Lightning-link Rapid technology can be used to label small quantities of antibody with 100% recovery
  • Type B biotin kit is intended for assays in which the biotinylated protein is captured by streptavidin immobilized on a surface (plates, nitrocellulose, magnetic beads, etc.)