Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ligroine, Hexane (Laboratory), Fisher Chemical

Catalog Number o338620
Click to view available options
Quantity:
20 L
Packaging:
Steel Pail
92112-69-1
C6H14
86.18
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
8058
CHEBI:29021
hexane
CCCCCC
92112-69-1
C6H14
86.18
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
8058
CHEBI:29021
hexane
CCCCCC
Melting Point -95°C
Color Colorless
Density 0.659g/cm³
Boiling Point 69°C
Quantity 20 L
Identification Pass Test
Packaging Steel Pail
Physical Form Liquid
Vapor Pressure 160mbar at 20°C
Viscosity 0.31 mPa/s at 20°C
Formula Weight 86.18
Grade Laboratory
Chemical Name or Material Ligroine, Hexane (Laboratory)
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Extremely flammable liquid and vapor. Irritating to eyes and skin. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of impaired fertility. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:3
Instability:0
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit