Learn More
Abnova™ LILRA3 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00011026T01
Catalog No. 89-950-678
Add to cart
Description
Description
Specifications
Specifications
293T
NM_006865.2
pCMV-LILRA3 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
LILRA3 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 48.4kDa
Transient overexpression cell lysate was tested with anti-LILRA3 antibody (Mfr. No. H00011026-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human