Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIM Kinase 1 Mouse anti-Human, Mouse, Rat, Monkey, PE/Atto594, Clone: OTI3H6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271141PEATT594

Catalog No. N271141P594


Add to cart

Description

Description

LIM Kinase 1 Monoclonal antibody specifically detects LIM Kinase 1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

LIM Kinase 1
OTI3H6
Flow Cytometry
Affinity Purified, Conjugated
LIMK1
Full length recombinant protein of human LIM Kinase 1 (NP_002305) produced in HEK293T cells.
Immunogen affinity purified
Breast Cancer, Cancer, Phospho Specific, Protein Kinase
Primary
3984.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
EC 2.7.11.1, LIM domain kinase 1, LIM motif-containing protein kinase, LIMKLIMK-1
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Monkey, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit