Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIM1 Mouse anti-Human, PE/Atto594, Clone: OTI2D5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271134PEATT594

Catalog No. N271134P594


Add to cart

Description

Description

LIM1 Monoclonal antibody specifically detects LIM1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

LIM1
OTI2D5
Flow Cytometry
Affinity Purified, Conjugated
LHX1
Recombinant protein fragment corresponding to amino acids 100-362 of human LIM1 (NP_005559) produced in HEK293T cells.
Immunogen affinity purified
DNA Repair, DNA replication Transcription Translation and Splicing, Neuroscience
Primary
3975.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
hLim-1, Homeobox protein Lim-1, LIM homeobox 1, LIM homeobox protein 1, LIM-1LIM/homeobox protein Lhx1, LIM1MGC138141, MGC126723
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit