Learn More
LIMA1 Mouse anti-Human, Clone: OTI1B4, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808565
Catalog No. 50-166-8945
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.
Specifications
Specifications
LIMA1
Monoclonal
Unconjugated
LIMA1
Lyophilized
LIMA1
Human recombit protein fragment corresponding to amino acids 1-275 of human LIMA1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
OTI1B4
PBS with 8% trehalose and no preservative; pH 7.3
Q9UHB6
EPLIN, SREBP3
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
51474