Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIMD1 Mouse anti-Human, Janelia Fluor 549, Clone: 3F2/G6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP250476JF549

Catalog No. NBP50476549


Add to cart

Description

Description

LIMD1 Monoclonal antibody specifically detects LIMD1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

LIMD1
3F2/G6
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
LIMD1
Recombinant LIMD1
Protein G purified
Store at 4°C in the dark.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
LIM domain-containing protein 1, LIM domains containing 1
Mouse
IgG1
0.1mL
Primary
8994
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit