Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIME Mouse anti-Mouse, Alexa Fluor 405, Clone: mLIME-05, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147230AF405

Catalog No. NBP147230AF


Add to cart

Description

Description

LIME Monoclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

LIME
mLIME-05
Western Blot
Purified
LIME1
Recombinant protein corresponding to amino acids 86-202 of mouse LIME
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Mouse monoclonal antibody mLIME-05 reacts with the cytoplasmic domain of LIME, a 30 kDa Lck-interacting transmembrane adaptor expressed mainly by T cells.
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
dJ583P15.4, FLJ20406, Lck interacting transmembrane adaptor 1, Lck-interacting membrane protein, Lck-interacting molecule, lck-interacting transmembrane adapter 1, LIMELP8067
Mouse
IgG2a
0.1 ml
Signal Transduction
Primary
54923
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit