Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lime Water, Saturated Unfiltered, Reagents

Lime Water, Saturated Unfiltered, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc C22325004A

Quantity 4 L

Catalog No. 01-337-195


Add to Cart

Description

Description

Specifications

Chemical Identifiers

7732-18-5 , 1305-62-0
18.02
water
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
O

Specifications

Colorless to Hazy-White
100°C
H2O
O
18.02
1.003
4 L
Liquid
7732-18-5 , 1305-62-0
0°C
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Poly Bottle
12.4
Non-sterile
Lime Water, Saturated Unfiltered
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.