Learn More
Limitin/IFN-zeta Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Manufacturer:  R&D Systems AF597SP
Catalog No. AF597SP
Add to cart
This item is not returnable. View return policy
Description
Description
Limitin/IFN-zeta Polyclonal antibody AF597SP specifically detects Limitin/IFN-zeta in Mouse samples. It is validated for Immunohistochemistry, Neutralization, Western Blotting
Specifications
Specifications
Limitin/IFN-zeta
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 1.5-7.5 ng/mL
Q9R1T0
6030405N23Rik, BGIF, Ifnz, IFN-z, IFNzeta, interferon zeta, Lmtn
Goat
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects Limitin/IFN-zeta in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse IFN-gamma is observed.
Mouse
Western Blot, Immunohistochemistry, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Ifnz
Mouse myeloma cell line NS0-derived recombinant mouse Limitin/IFN-zeta Leu22-Arg182 Accession # Q9R1T0
Immunogen affinity purified
RUO
Primary
319146
Reconstitute at 0.2 mg/mL in sterile PBS.