Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Limitin/IFN-zeta Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0123-7D19, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212426AF750

Catalog No. NP212426A70


Add to cart

Description

Description

Limitin/IFN-zeta Monoclonal antibody specifically detects Limitin/IFN-zeta in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Limitin/IFN-zeta
RM0123-7D19
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
Ifnz
Mouse
Protein G purified
Store at 4°C in the dark.
Monoclonal
Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
6030405N23Rik, BGIF, Ifnz, interferon zeta, Lmtn
Rat
IgG2
0.1 mL
Primary
319146.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit