Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LIMS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563557
Description
Immunogen sequence: KRLKKLSELT SRKAQPKATD LNSA Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 71%.
Adapter protein in a cytoplasmic complex linking beta- integrins to the actin cytoskeleton, bridges the complex to cell surface receptor tyrosine kinases and growth factor receptors. Plays a role in modulating cell spreading and migration.
Specifications
LIMS2 | |
Polyclonal | |
Unconjugated | |
Lims2 | |
ILK-binding protein; LGMD2W; LIM and senescent cell antigen like domains 2; LIM and senescent cell antigen-like domains 2; LIM and senescent cell antigen-like-containing domain protein 2; LIM zinc finger domain containing 2; LIM-like protein 2; Lims2; LIM-type zinc finger domains 2; Particularly interesting new Cys-His protein 2; Pinch2; PINCH-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55679 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z4I7 | |
Lims2 | |
Recombinant protein corresponding to Human LIMS2. Recombinant protein control fragment (Product #RP-101844). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction