Learn More
FUJIFILM Wako Limulus Color KY Multi-Test
Quantitative kinetic chromogenic assays (KCA) that are endotoxin-specific and unreactive to (1,3)-β-D-glucan, multi-test vials for use with Toxinometer™ or microplate reader, ideal for user with larger number of samples that are turbid
Manufacturer:  FUJIFILM Wako 29153101
Catalog No. 12-333-026
Add to cart
Description
Description
  • Reagent reacts with endotoxin over wide range of concentrations
  • Reduces chance of interfering factors due to high sensitivity
  • Can be used with Toxinometer™ or microplate reader
  • KCA quantitative range detection limit of 0.0005 to 5 EU/mL
  • Sample size is 0.1 mL when used with tube reader
  • 0.05 mL sample size when used with microplate reader
Specifications
Specifications
Lyophilized Powder
2 mL
For use with Toxinometer™ or microplate reader
Reagent
3 x 2 mL vials of lysate and 1 vial of CSE (500 ng/vial)