Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIN-28A Mouse anti-Human, HRP, Clone: 1086, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243673H

Catalog No. NBP243673H


Add to cart

Description

Description

LIN-28A Monoclonal antibody specifically detects LIN-28A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

LIN-28A
1086
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
LIN28A
Recombinant protein encompassing a sequence within the center region of human LIN28. The exact sequence is proprietary.
Protein G purified
Embryonic Stem Cell Markers, Epigenetics, microRNA, Neuroscience, Stem Cell Markers, Stem Cells
Primary
79727
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
HRP
PBS with No Preservative
CSDD1lin-28A, FLJ12457, LIN-28, lin-28 homolog (C. elegans), lin-28 homolog A (C. elegans), Lin-28A, LIN28RNA-binding protein LIN-28, ZCCHC1protein lin-28 homolog A, Zinc finger CCHC domain-containing protein 1, zinc finger, CCHC domain containing 1
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit