Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIN-28B Sheep anti-Human, Polyclonal, R&D Systems
Sheep Polyclonal Antibody
Manufacturer: R&D Systems AF7726SP
This item is not returnable.
View return policy
Description
LIN-28B Polyclonal antibody AF7726SP specifically detects LIN-28B in Human samples. It is validated for Immunohistochemistry, Western BlottingSpecifications
LIN-28B | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Q6ZN17 | |
CSDD2, CSDD2protein lin-28 homolog B, FLJ16517, lin-28 homolog B (C. elegans), Lin-28.2, lin-28B, LIN28B | |
Sheep | |
IgG | |
25 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Polyclonal | |
Detects human LIN-28B in direct ELISAs and Western blots. In direct ELISAs, less than 4% cross-reactivity with recombinant human LIN-28A is observed. | |
Human |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Affinity Purified | |
LIN28B | |
E. coli-derived recombinant human LIN-28B Val83-Thr250 Accession # Q6ZN17 | |
Immunogen affinity purified | |
RUO | |
Primary | |
389421 | |
Sterile PBS to a final concentration of 0.2 mg/mL. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only