Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIN9 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | LIN9 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179424
|
Novus Biologicals
NBP179424 |
100 μL |
Each for $436.00
|
|
NBP17942420
|
Novus Biologicals
NBP17942420UL |
20 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
LIN9 Polyclonal specifically detects LIN9 in Human samples. It is validated for Western Blot.Specifications
LIN9 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
BARA, BARPsv, hLin-9, huLin-9, Lin-9, lin-9 homolog (C. elegans), pRB-associated protein, protein lin-9 homolog, rb related pathway actor, TGS1, TGSBeta subunit-associated regulator of apoptosis, TUDOR gene similar protein, Type I interferon receptor beta chain-associated protein | |
LIN9 | |
IgG | |
Affinity Purified | |
64 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_775106 | |
286826 | |
Synthetic peptide directed towards the N terminal of human LIN9The immunogen for this antibody is LIN9. Peptide sequence SLQKTPVWKGRNTSSAVEMPFRNSKRSRLFSDEDDRQINTRSPKRNQRVA. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title