Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Linalool 97%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
2.5L
Packaging:
Glass bottle
78-70-6
C10H18O
154.25
MFCD00008906
CDOSHBSSFJOMGT-UHFFFAOYSA-N
linalool,linalol,3,7-dimethyl-1,6-octadien-3-ol,allo-ocimenol,beta-linalool,linalyl alcohol,1,6-octadien-3-ol, 3,7-dimethyl,+--linalool,p-linalool,linanool
6549
CHEBI:17580
3,7-dimethylocta-1,6-dien-3-ol
CC(=CCCC(C)(C=C)O)C
78-70-6
C10H18O
154.25
MFCD00008906
CDOSHBSSFJOMGT-UHFFFAOYSA-N
linalool,linalol,3,7-dimethyl-1,6-octadien-3-ol,allo-ocimenol,beta-linalool,linalyl alcohol,1,6-octadien-3-ol, 3,7-dimethyl,+--linalool,p-linalool,linanool
6549
CHEBI:17580
3,7-dimethylocta-1,6-dien-3-ol
CC(=CCCC(C)(C=C)O)C
Density 0.8610g/mL
Viscosity 4.4 mPa.s (25°C)
Color Colorless to Yellow
Infrared Spectrum Authentic
Flash Point 75°C
Refractive Index 1.4600 to 1.4640
Specific Gravity 0.861
Quantity 500mL
Specific Rotation -2° to +2° (20°C, 589nm) (neat)
Packaging Glass bottle
Boiling Point 199.0°C
Assay Percent Range 97%
Linear Formula (CH3)2C=CHCH2CH2C(CH3)(OH)CH=CH2
Beilstein 01, 462
Merck Index 15, 5550
Solubility Information Solubility in water: 1.45g/L (25°C). Other solubilities: miscible with alcohol and ether
Formula Weight 154.25
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Linalool, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 201-134-4

RTECSNumber : RG5775000

TSCA : TSCA