Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lindberg/Blue M™ Vacuum Ovens

Benefit from a digital control, built-in over-temperature protection and a flexible vacuum/purge/release system for a range of applications with these ovens.

Manufacturer:  Thermo Scientific™ VO1218C

 View more versions of this product

Catalog No. 10-549-224


  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Add to cart

Alert:

Ovens are not suitable for use with hazardous vapor. For use with nonflammable, noncorrosive inert gases only.


Specifications

Description

Benefit from a digital control, built-in over-temperature protection and a flexible vacuum/purge/release system with Thermo Scientific™ Lindberg/Blue M™ Vacuum Ovens. These ovens work well with a range of applications including drying, curing, outgassing, aging, process control and other applications requiring elevated temperature in reduced atmospheres or vacuum/purge with nonflammable and inert atmospheres.

Single-Setpoint Digital Microprocessor Control

  • Maximum temperature: 260°C (500°F) with nominal uniformity of ±3.5% of setpoint
  • Simultaneous LED display of setpoint and actual temperature with push-button view; resolution to within 1°C
  • Keypad data entry and pull-out control module for easy service
  • Independent overtemperature safety system factory set with reset pushbutton

Vacuum System

  • 1in.(2.54cm) manifold, chamber rear, exterior, connects to pump or in-house vacuum source
  • With inert gas injection valve, fresh air inlet, vacuum exhaust port, controllable vacuum release vent port
  • 1000 microns (1 torr) capacity
  • Vacuum gauge, vacuum, gas and inlet valves located on front

Construction

  • Corrosion-resistant stainless-steel cabinet includes three aluminum shelves with painted or stainless-steel finish
  • Rear-mounted vacuum manifold connects to pump (not included) or in-house vacuum source
  • Low-watt heating element extends oven life
  • Heavy-gauge steel, dual-wall construction with fiberglass insulation for minimum heat loss in walls and door
  • High-temperature silicone gasket is specially molded for a positive seal
  • Magnetic latch for positive door seal (4.5 cu. ft. capacity models include mechanical latches)
  • Viewing windows: 0.5in.(1.3cm) thick in two smaller capacity models or 0.75in.(1.9cm) thick in 4.5 cu. ft. model; permit observation of work in progress without loss of heat
  • Stainless-steel exterior models come with double door—a solid outer door over inner window door

Includes:

Power cord and aluminum shelves (number of shelves based on model; see ordering table)

Description & Specifications

Specifications

1.5 cu.ft., 42.5L
18 x 12 x 12 in. (46 x 31 x 31cm)
208 to 240V 50/60Hz
42.5L; 208/240V; 1250w; Painted cabinet; 1 door, 3 shelves
42.5L
18 in.
LED
12 in.
31cm
26 in.
Inert gas port, vacuum port and vent port
3
113kg
6° to 260°C
±3.5°C
208 to 240V
21 in.
53cm
42.5L; 208/240V; 1250w; Painted cabinet; 1 door, 3 shelves
26 x 21 x 29 in. (66 x 53 x 74cm)
Painted Steel
1.5 cu.ft.
Corrosion-resistant stainless steel
46cm
29 in.
74cm
50/60Hz
66cm
Overtemperature Safety System
250 lb.
Digital Microprocessor Control
±1°C
1 x 10-2 torr (10 microns)
1250w
12 in.
31cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit