Learn More
Novus Biologicals™ LINGO3 Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP255062PEP
Catalog No. NBP55062PEP
Add to cart
Description
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human LINGO3. The LINGO3 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
Antibody Competition
645191
>80% by SDS-PAGE and Coomassie blue staining
E. coli
RUO
LINGO3
PBS and 1M Urea, pH 7.4.
26 kDa
100 μl
Store at -20°C. Avoid freeze-thaw cycles.
LINGO3
Human