Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cliniqa™ LiniCAL Cardiac Calibration Verifier for Ortho Systems

LiniCAL Cardiac Calibration Verifier for Ortho Systems is convenient and cost effective. Five levels include NT-proBNP, CKMB, Myoglobin and cTnI to allow monitoring of the reportable range. Kit includes two vials per level for multiple testing events, and has closed and open vial stability.

Manufacturer:  Cliniqa™ 98750

Catalog No. 22-100-108


Due to product restrictions, please to purchase or view availability for this product.

This item is not returnable. View return policy

Description

Description

Cliniqa™ LiniCAL Cardiac Calibration Verifier for Ortho Systems is a convenient liquid stable human-serum matrix that is ready-to-use and store at 2 to 8°C. Two events of five levels of assayed materials are designed for use with Ortho platforms in order to challenge the low and high limits of the analyzer's reportable range. Analytes included are NT-proBNP, CKMB, Myoglobin and cTnI. LiniCAL sets meet CLIA and state regulatory reporting for 3rd party calibration verification while also being very useful for troubleshooting after preventive maintenance or a new installation.

  • Liquid 2 to 8°C ready-to-use for easy storage and long-term performance
  • Two vials per level to accommodate multiple test events
  • Five levels to cover a wide range of normal and abnormal results
  • Comprehensive list of the most commonly tested constituents
  • Excellent closed and open vial stability
Specifications

Specifications

2 years
Human source
Open vial: 30 days (at 2°C to 8°C)
3 mL
Yes
NT-proBNP, CKMB, Myoglobin and cTnI
Refrigerated at 2°C to 8°C
Calibration Verifier
Liquid
5 levels
Human Serum
No
Ortho Systems
Ortho Systems
Kit Box (5.31 x 4.06 x 3.125 in.)
Cold Packs, at 2°C to 8°C
Safety and Handling

Safety and Handling

SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit