Learn More
Cliniqa™ LiniCAL Protein 2 Calibration Verifier for Beckman Systems
LiniCAL Protein 2 Calibration Verifier for Beckman Systems is convenient and cost effective. Five levels include ASO, CRP, C3, C4, FER, IgA, IgG, IgM, PAB and TRF for monitoring of the reportable range. Includes two vials per level for multiple testing events, and has closed and open vial stability.
Supplier: Cliniqa™ 98302
Description
Cliniqa™ LiniCAL Protein 2 Calibration Verifier for Beckman Systems is a convenient liquid stable human-serum matrix that is ready-to-use and store at 2 to 8°C. Two events of five levels of assayed materials are designed for use with Beckman platforms in order to challenge the low and high limits of the analyzer's reportable range. Analytes included are ASO, CRP, C3, C4, FER, IgA, IgG, IgM, PAB and TRF. LiniCAL sets meet CLIA and state regulatory reporting for 3rd party calibration verification while also being very useful for troubleshooting after preventive maintenance or a new installation.
- Liquid 2 to 8°C ready-to-use for easy storage and long-term performance
- Two vials per level to accommodate multiple test events
- Five levels to cover a wide range of normal and abnormal results
- Comprehensive list of the most commonly tested constituents
- Excellent closed and open vial stability
Specifications
Liquid 2°C to 8°C | |
Antistreptolysin O (ASO), C-Reactive Protein (CRP), C3, C4, Ferritin, IgA, IgG, IgM, PAB, Transferrin (TRF) | |
Beckman Coulter Systems - Clinical Invitro Diagnostic | |
Yes | |
No | |
Kit Box (5.31 x 4.06 x 3.125 in.) | |
Human Serum Base | |
Liquid |
3 mL fill | |
Calibration | |
Calibration Verifier | |
5 levels | |
Human Serum | |
Cold Packs, at 2°C to 8°C | |
14 days (at 2°C to 8°C) |
Safety and Handling
ShelfLife : 3 years
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.