Learn More
Cliniqa™ LiniCAL Protein 2 Calibration Verifier for Roche Systems
LiniCAL Protein 2 Calibration Verifier for Roche Systems is convenient and cost effective. Five levels include AAG, AAT, CER and B2M to allow monitoring of the reportable range. Kit includes two vials per level for multiple testing events, and has closed and open vial stability.
Supplier: Cliniqa™ 98502
Description
Cliniqa™ LiniCAL Protein 2 Calibration Verifier for Roche Systems is a convenient liquid stable human-serum matrix that is ready-to-use and store at 2 to 8°C. Two events of five levels of assayed materials are designed for use with Roche platforms in order to challenge the low and high limits of the analyzer's reportable range. Analytes included are AAG, AAT, CER and B2M. LiniCAL sets meet CLIA and state regulatory reporting for 3rd party calibration verification while also being very useful for troubleshooting after preventive maintenance or a new installation.
- Liquid 2 to 8°C ready-to-use for easy storage and long-term performance
- Two vials per level to accommodate multiple test events
- Five levels to cover a wide range of normal and abnormal results
- Comprehensive list of the most commonly tested constituents
- Excellent closed and open vial stability
Specifications
Refrigerated at 2°C to 8°C | |
AAG, AAT, Ceruloplasmin, beta-2-microglobulin (B2M) | |
Calibration Verifier | |
5 levels | |
Human Serum | |
Cold Packs, at 2°C to 8°C | |
Open vial: 30 days (at 2°C to 8°C) |
1 mL fill | |
Roche Systems | |
Yes | |
No | |
Kit Box (5.31 x 4.06 x 3.125 in.) | |
Human Serum | |
Liquid |
Safety and Handling
ShelfLife : 3 years
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.