Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cliniqa™ LiniCAL Protein 4 Calibration Verifier for Roche Systems

LiniCAL Protein 4 Calibration Verifier for Roche Systems is convenient and cost effective. Six levels include ALB, PREA and RF to allow monitoring of the reportable range. Kit includes two vials per level for multiple testing events, and has closed and open vial stability.

Manufacturer:  Cliniqa™ 98504

Catalog No. 22-100-112


Due to product restrictions, please to purchase or view availability for this product.

This item is not returnable. View return policy

Description

Description

Cliniqa™ LiniCAL Protein 4 Calibration Verifier for Roche Systems is a convenient liquid stable human-serum matrix that is ready-to-use and store at 2 to 8°C. Two events of six levels of assayed materials are designed for use with Roche platforms in order to challenge the low and high limits of the analyzer's reportable range. Analytes included are ALB, PREA and RF. LiniCAL sets meet CLIA and state regulatory reporting for 3rd party calibration verification while also being very useful for troubleshooting after preventive maintenance or a new installation.

  • Liquid 2 to 8°C ready-to-use for easy storage and long-term performance
  • Two vials per level to accommodate multiple test events
  • Six levels to cover a wide range of normal and abnormal results
  • Comprehensive list of the most commonly tested constituents
  • Excellent closed and open vial stability
Specifications

Specifications

Liquid
Human serum base
30 days (at 2°C to 8°C)
1 mL
Yes
Roche Systems
Kit Box (5.31 x 4.06 x 3.125 in.)
Cold Packs, at 2°C to 8°C
3 years
6 Levels
Human Serum
No
ALB, PREA and RF
Refrigerated at 2°C to 8°C
Calibration Verifier
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit