Learn More
Selleck Chemical LLCSupplier Diversity Partner Linifanib (ABT-869) 10mg 796967-16-3 AL39324,RG3635  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S100310MG
Linifanib (ABT-869, AL39324, RG3635) is a novel, potent ATP-competitive VEGFR/PDGFR inhibitor for KDR, CSF-1R, Flt-1/3 and PDGFRβ with IC50 of 4 nM, 3 nM, 3 nM/4 nM and 66 nM respectively, mostly effective in mutant kinase-dependent cancer cells (i.e. FLT3). Linifanib (ABT-869) induces autophagy and apoptosis. Phase 3.
Catalog No. 50-686-8