Learn More
Linoleic acid, 95%, Thermo Scientific Chemicals
Description
Unsaturated μ-6 fatty acid Linoleic acid is involved in the production of quick-drying oils which are used in oil paints and varnishes. It is also used in beauty products due to its beneficial properties on the skin such as moisture retention. It is a key starting material for the synthesis of linoleyl alcohol and octadecanoic acid. It is used as a precursor of arachidonic acid (AA) and various prostaglandins. It may be used to improve the delivery and efficacy of anti-cancer drugs and in cancer protection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Unsaturated µ-6 fatty acid Linoleic acid is involved in the production of quick-drying oils which are used in oil paints and varnishes. It is also used in beauty products due to its beneficial properties on the skin such as moisture retention. It is a key starting material for the synthesis of linoleyl alcohol and octadecanoic acid. It is used as a precursor of arachidonic acid (AA) and various prostaglandins. It may be used to improve the delivery and efficacy of anti-cancer drugs and in cancer protection.
Solubility
Miscible with dimethyl sulfoxide, ethanol, ether, petroleum ether, dimethylformamide, acetone and benzene. Insoluble in water.
Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | -12°C |
Density | 0.902 |
Boiling Point | 230°C (16 mmHg) |
Flash Point | >110°C (230°F) |
Odor | Weak, Characteristic |
Refractive Index | 1.4699 |
Linear Formula | CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H |
Quantity | 5 g |
Beilstein | 1727101 |
Sensitivity | Air sensitive |
Show More |
Safety and Handling
EINECSNumber | 200-470-9 |
RTECSNumber | RF9990000 |
TSCA | Yes |
Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.