Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LINS1 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30392520UL

 View more versions of this product

Catalog No. NB094889


Add to cart

Description

Description

LINS1 Polyclonal antibody specifically detects LINS1 in Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

LINS1
Unconjugated
Purified
Rabbit
IgG
20 μL
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Mouse, Rat
Western Blot
PBS with 50% glycerol, pH7.3.
FLJ10583, lines homolog (Drosophila), lines homolog 1, LINS1, protein Lines homolog, protein Lines homolog 1, WINS1 protein with Drosophila Lines (Lin) homologous domain, wnt-signaling molecule Lines homolog 1
A synthetic peptide corresponding to a sequence within amino acids 1-100 of mouse LINS1 (NP_690028.2).
Affinity purified
RUO
Primary
55180
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit