Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LINS1 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen LINS1
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094888
View Documents
Novus Biologicals
NBP303925100UL
100 μg Each for $320.71
Add to cart
 
NB094889
View Documents
Novus Biologicals
NBP30392520UL
20 μg Each for $138.37
Add to cart
 
Description

Description

LINS1 Polyclonal antibody specifically detects LINS1 in Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

LINS1
Unconjugated
Rabbit
Mouse, Rat
FLJ10583, lines homolog (Drosophila), lines homolog 1, LINS1, protein Lines homolog, protein Lines homolog 1, WINS1 protein with Drosophila Lines (Lin) homologous domain, wnt-signaling molecule Lines homolog 1
A synthetic peptide corresponding to a sequence within amino acids 1-100 of mouse LINS1 (NP_690028.2).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
55180
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit