Learn More
LION™ MT94™ Multi-Threat CBRN Garment
CBRN garment provides vapor, liquid, and FR protection for chem/bio incidents and is certified to meet NFPA 1994/Class 2 and NFPA 1992
Manufacturer:  LION™ CMTM10XL
 View more versions of this product
Catalog No. 19-813-183
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Rugged GORE™ CHEMPAK™ Ultra Barrier fabric features thin, lightweight, and high-strength PTFE film sandwiched within tough DuPont™ Nomex™ outer and inner layers
  • Streamlined design and light weight deliver optimum comfort and natural freedom of movement
  • Rugged construction allows you to perform in the most challenging of circumstances
  • Re-usable for multiple wears and washings, if not exposed or contaminated
  • Certified for use with Gore G9492 gloves and OnGuard HAZMAX™ 87012 Boots
  • Hook and loop belt loops to hold equipment belt securely
  • ID Velcro™ patches on chest and shoulders for hook and loop mounted badges
  • Roomy thigh cargo pockets
  • Hook and loop wrist and leg cuff adjustments for secure fit
  • O-ring inner glove
  • Avon molded face seal
  • NTOA Member tested and recommended
  • Navy blue
Technical rescue, Contaminated rescue, Decon, SWAT, HAZMAT, Confined space rescue, Explosives Ordnance Disposal
Certifications
Certified to NFPA 1994 and 1992 Class 2, Current Edition
Specifications
Specifications
Hook and loop
X-Large