Learn More
Agilent BioTek Lionheart FX Automated Microscope, IVD
Lionheart FX automated microscope is a compact system for a broad range of imaging workflows. For In Vitro Diagnostic Use.
Supplier:  Agilent BioTek LFXWSI
Request Bulk Order
Catalog No. BTLFXWSI
Item Discontinued This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or send email to cs.webemail@thermofisher.com to discuss your specific assay requirements.
This item is not returnable. View return policy
Description
Description
Lionheart FX automated microscope is a compact system for a broad range of imaging workflows. It offers up to 60x air; 60x and 100x oil immersion magnification, with fluorescence, brightfield, color brightfield, and phase contrast channels for maximum application reach
An optional environment control cover provides incubation to 40°C and effective containment for CO2/O2 control. A humidity chamber optimizes conditions for long-term live cell imaging applications, and an available dual reagent injector facilitates rapid kinetic assays.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cell Imager
Live Cell Imaging
High Power LED
18.3 in.
14 in.
17.9 in.
CE and TUV marked. RoHS compliant.
50/60 Hz
CCD
LFX
Gen5 Microplate Reader and Imager
46.5 cm
35.6 cm
45.5 cm
1 Year
110/220 V