Learn More
Agilent BioTek Lionheart FX Automated Microscope, IVD
Lionheart FX automated microscope is a compact system for a broad range of imaging workflows. For In Vitro Diagnostic Use.
Supplier: Agilent BioTek LFXSI
Description
Lionheart FX automated microscope is a compact system for a broad range of imaging workflows. It offers up to 60x air; 60x and 100x oil immersion magnification, with fluorescence, brightfield, color brightfield, and phase contrast channels for maximum application reach
An optional environment control cover provides incubation to 40 °C and effective containment for CO2/O2 control. A humidity chamber optimizes conditions for long-term live cell imaging applications, and an available dual reagent injector facilitates rapid kinetic assays.
Specifications
Cell Imager | |
Live Cell Imaging | |
High Power LED | |
18.3 in. | |
14 in. | |
17.9 in. | |
CE and TUV marked. RoHS compliant. | |
50/60 Hz |
16-bit Gray Scale, Sony CCD, 1.25 Megapixel | |
LFX | |
Gen5 Microplate Reader and Imager | |
46.5 cm | |
35.6 cm | |
45.5 cm | |
1 year | |
110/220 V |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.