Learn More
Agilent BioTek Lionheart LX Automated Microscope, IVD
Lionheart LX automated microscope is a compact system for a broad range of imaging workflows. For In Vitro Diagnostic Use.
Supplier:  Agilent BioTek LLXWSI
Catalog No. BTLLXWSI
Item Discontinued This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or send email to cs.webemail@thermofisher.com to discuss your specific assay requirements. View substitute products »
This item is not returnable. View return policy
Description
Description
The Lionheart LX automated microscope is designed for affordability, offering an alternative to expensive, custom-built automated microscopes that are complex to learn and use. The compact, ocular free hardware design is more comfortable to use and reduces ergonomic distress in operators
Along with Gen5 software, Lionheart LX enables Augmented Microscopy, which fully automates image capture, processing, and analysis. Operation is easy, load samples, start a run and gather publication-ready images and quantitative data. Lionheart LX can increase your research output and reduce time and costs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cell Imager
Cell Counting
Gen5 3.0 Microplate Reader and Imager Software included; Gen5 Image+ and Image Prime software for advanced image analysis (option)
46.5 cm
35.6 cm
17.9 in.
CE and TUV marked. RoHS compliant.
50/60 Hz
CMOS
LLX
18.3 in.
14 in.
51 kg
45.5 cm
1 Year
110/220 V