Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioTek Instruments Lionheart LX Automated Microscope Greener Choice Available on GSA/VA Contract for Federal Government customers only.

BioTek Instruments Lionheart LX Automated Microscope is affordable automated digital microscopy for microplates, slides, cell culture dishes and flasks

Manufacturer:  BioTek Instruments LLX

Catalog No. BTLLX


Add to cart

Description

Description

  • High contrast brightfield, color brightfield and fluorescence imaging
  • Augmented Microscopy with seamless image capture
  • Quick Analyze feature: instant cell count and confluence measurement

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Warranty and Services

12 months

Specifications

Specifications

50/60 Hz
16-bit gray scale, Sony CMOS, 1.25 megapixel
12 months
16-bit gray scale, Sony CMOS, 1.25 megapixel
110/220 V
14 in.
60 W
17.9 in.
Lionheart LX Automated Microscope.
Imaging
50/60 Hz
18.3 in.
Cell counting, end point live cell assay, H&E, label free cell counting, confluence, genotoxicity, immunofluorescence, NASH, Phenotypic
Gen5™ 3.0 Microplate Reader and Imager Software included;Gen5 Image+ and Image Prime software for advanced image analysis (option)
51 kg
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit