Learn More
BioVision IncSupplier Diversity Partner Lipase Activity Colorimetric Assay Kit: High-Throughput Suitable Colorimetric Assay for Measuring Lipase Activity in various biological Samples such as Serum, Tissues, Cells etc. Simple, Sensitive & Accurate. Detection Limit: 0.02 mU. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K722100
Lipases perform essential roles in the digestion, transport and processing of dietary lipids (e.g. fats and oils) in living organisms. In humans, pancreatic lipase is the key enzyme responsible for breaking down fats in the digestive system by converting triglyceride to monoglyceride and free fatty acid. Pancreatic lipase monitoring is also used to help diagnose Crohn's disease, cystic fibrosis and celiac disease. Damage to the pancreas can exhibit a 5-10 fold increase of serum lipase levels within 24 to 48 hours. In BioVision’s Lipase Assay Kit, lipase hydrolyzes a triglyceride substrate to form glycerol which is quantified enzymatically by via monitoring a linked change in the OxiRed probe absorbance (λ=570nm). This assay is rapid, simple, sensitive, and reliable, as well as, suitable for high throughput activity screening of lipase. This kit detects lipase activity as low as 10mU per well. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. NC0123297