Learn More
BioVision IncSupplier Diversity Partner Lipase Activity Colorimetric Assay Kit II: High-Throughput Suitable Colorimetric Assay for Measuring Lipase Activity in various biological Samples such as Serum, Tissues, Cells etc. Simple, Sensitive & Accurate. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K723100
Lipase is a subclass of the esterases that catalyze the hydrolysis of ester bonds in water-insoluble, lipid substrates. Lipases perform essential roles in the digestion, transport and processing of dietary lipids (e.g. triglycerides, fats, oils) in most, if not all, living organisms. In humans, pancreatic lipases are the key enzyme responsible for breaking down fats in the digestive system by converting triglycerides to monoglycerides and free fatty acids. During the damage of the pancreas, lipase levels can rise 5 to 10-fold within 24 to 48 hours. The kit provides a simple, sensitive, and reliable assay for rapid analysis of Lipase in samples. In the assay, Lipases hydrolyze a specific substrate to generate a product which reacts with the DTNB probe to generate color (λ=412 nm). The kit is also suitable for high throughput analyses. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9859