Learn More
Cell Biolabs IncSupplier Diversity Partner LIPID EXTRACTION KIT TRL SIZE  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA612T
The Lipid Extraction Kit (Chloroform-Free) uses a chloroform-free organic system that results in the organic phase being on the top, making it easier to use while eliminating the need for chloroform. 10 preps/trial size kit.
Catalog No. 50-673-654