Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lipin 3 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10053802
Description
Lipin 3 Polyclonal specifically detects Lipin 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
Lipin 3 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
dJ450M14.2, dJ450M14.3, dJ620E11.2, EC 3.1.3.4, lipin 3, lipin 3-like, lipin-3, Lipin-3-like, LIPN3L, phosphatidate phosphatase LPIN3, SMP2 | |
Goat | |
Affinity Purified | |
RUO | |
64900 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Immunohistochemistry 2 - 3 ug/ml, Immunohistochemistry-Paraffin 2 - 3 ug/ml, Peptide ELISA Detection limit 1:64000 | |
NP_075047.1 | |
LPIN3 | |
Peptide with sequence C-KPKQKEDAVATD corresponding to internal region according to NP_075047.1. | |
0.1 mg | |
Primary | |
Cross reacts with Human. Expected from sequence similarity: Dog and Pig. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction