Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipocalin-2/NGAL Mouse anti-Canine, Human, Mouse, Rat, Alexa Fluor™ 750, Clone: 14, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP241364AF750

Catalog No. NP241364A70


Add to cart

Description

Description

Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Canine, Human (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Sandwich ELISA.
Specifications

Specifications

Lipocalin-2/NGAL
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
LCN2
Recombinant dog NGAL adsorbed onto aluminum hydroxide gel
100 μL
Primary
Antibody binds dog NGAL
Store at 4°C in the dark.
Western Blot, ELISA, Immunohistochemistry
14
Western Blot, ELISA, Immunohistochemistry, Sandwich ELISA
24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin
Mouse
Protein A or G purified
Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuroscience
3934.0
Canine, Human, Mouse, Rat
IgG2b κ
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit