Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipocalin-2/NGAL Mouse anti-Human, Alexa Fluor 488, Clone: 15, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP130003AF488

Catalog No. NBP130003AG


Add to cart

Description

Description

Lipocalin-2/NGAL Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications

Specifications

Lipocalin-2/NGAL
15
Western Blot, ELISA
Purified
LCN2
Recombinant human NGAL adsorbed onto aluminum hydroxide gel
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin
Mouse
IgG1 Kappa
0.1 ml
Lipid and Metabolism
Primary
3934
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit