Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipocalin-2/NGAL Mouse anti-Human, Janelia Fluor 549, Clone: 5G5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223529JF549

Catalog No. NBP23529549


Add to cart

Description

Description

Lipocalin-2/NGAL Monoclonal antibody specifically detects Lipocalin-2/NGAL in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Lipocalin-2/NGAL
5G5
Western Blot, ELISA, Immunohistochemistry
Purified
LCN2
Human recombinant NGAL in monomer and dimer forms adsorbed to aluminum hydroxide gel
Protein A or G purified
Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuroscience
Primary
3934
Human
ELISA, Immunohistochemistry, Western Blot
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Clone 5G5 is specific for human NGAL. No cross-reaction with NGAL from other species has been detected.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit