Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipocalin-2/NGAL Mouse anti-Monkey, Alexa Fluor 532, Clone: 56, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP241366AF532

Catalog No. NP241366A32


Add to cart

Description

Description

Lipocalin-2/NGAL Monoclonal antibody specifically detects Lipocalin-2/NGAL in Monkey samples. It is validated for ELISA, Immunohistochemistry.
Specifications

Specifications

Lipocalin-2/NGAL
56
ELISA, Immunohistochemistry, Sandwich ELISA
Conjugated, Purified
LCN2
Recombinant monkey NGAL
Protein A or G purified
Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuroscience
Primary
3934.0
Monkey
ELISA, Immunohistochemistry
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Antibody reacts with NGAL (neutrophil gelatinase-associated lipocalin) from Macaca mulatta (Rhesus) and Macaca facicularis (Cynomolgus)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit